Assay Details
Gene Symbol: RSPO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001319025.1 | NP_001305954.1 | 2 - 3 | 284 | 76 | |
GenBank mRNA | CR406671.1 | - | 1 - 2 | 122 | 76 | |
KC250012.1 | - | 2 - 3 | 284 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
