Assay Details
| Gene Symbol: XIRP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204348.3 | NP_989679.3 | 4 | 3614 | 97 | |
| XM_015281203.2 | XP_015136689.1 | 8 | 4258 | 97 | ||
| GenBank mRNA | AF051944.2 | - | 4 | 3580 | 97 | |
| CR406895.1 | - | 1 | 943 | 97 | ||
More Information
Set Membership: |
|

XIRP1
Genomic Map