Assay Details
| Gene Symbol: MGMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025152041.1 | XP_025007809.1 | 6 | 1670 | 153 | |
| XM_025152042.1 | XP_025007810.1 | 6 | 1662 | 153 | ||
| XM_025152044.1 | XP_025007812.1 | 6 | 1511 | 153 | ||
| XM_025152045.1 | XP_025007813.1 | 6 | 1552 | 153 | ||
| GenBank mRNA | CR407154.1 | - | 1 | 747 | 153 | |
More Information
Set Membership: |
|

MGMT
Genomic Map