Assay Details
| Gene Symbol: SHROOM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004938410.3 | XP_004938467.3 | 9 - 10 | 4991 | 111 | |
| XM_025146614.1 | XP_025002382.1 | 9 - 10 | 5033 | 111 | ||
| XM_025146615.1 | XP_025002383.1 | 7 - 8 | 4669 | 111 | ||
| XM_025146616.1 | XP_025002384.1 | 5 - 6 | 1828 | 111 | ||
| GenBank mRNA | CR407446.1 | - | 1 - 2 | 161 | 111 | |
More Information
Set Membership: |
|

SHROOM2
Genomic Map