Assay Details
| Gene Symbol: PMP22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277071.2 | NP_001264000.1 | 2 - 3 | 221 | 59 | |
| NM_001277072.2 | NP_001264001.1 | 2 - 3 | 194 | 59 | ||
| GenBank mRNA | BU280570.1 | - | 2 - 3 | 169 | 59 | |
| BU342808.1 | - | 2 - 3 | 181 | 59 | ||
| BX935997.1 | - | 2 - 3 | 221 | 59 | ||
| CR523174.1 | - | 2 - 3 | 170 | 59 | ||
More Information
Set Membership: |
|

PMP22
Genomic Map