Assay Details
| Gene Symbol: SMAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001201455.1 | NP_001188384.1 | 2 - 3 | 657 | 60 | |
| XM_015276129.2 | XP_015131615.2 | 3 - 4 | 1014 | 60 | ||
| XM_015276130.2 | XP_015131616.2 | 3 - 4 | 1041 | 60 | ||
| XM_015276131.2 | XP_015131617.2 | 3 - 4 | 1227 | 60 | ||
| XM_015276132.2 | XP_015131618.2 | 3 - 4 | 907 | 60 | ||
| XM_015276134.2 | XP_015131620.1 | 2 - 3 | 2083 | 60 | ||
| XM_025149512.1 | XP_025005280.1 | 3 - 4 | 1188 | 60 | ||
| XM_025149513.1 | XP_025005281.1 | 8 - 9 | 1480 | 60 | ||
| XM_025149514.1 | XP_025005282.1 | 4 - 5 | 1122 | 60 | ||
| GenBank mRNA | AF143239.1 | - | 1 - 2 | 160 | 60 | |
| AF233238.1 | - | 2 - 3 | 613 | 60 | ||
| AY953143.1 | - | 2 - 3 | 657 | 60 | ||
More Information
Set Membership: |
|

SMAD1
Genomic Map