Assay Details
| Gene Symbol: DACH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199446.1 | NP_001186375.1 | 4 - 5 | 1156 | 65 | |
| XM_015275195.2 | XP_015130681.1 | 5 - 6 | 2030 | 65 | ||
| XM_015275204.2 | XP_015130690.1 | 5 - 6 | 2030 | 65 | ||
| XM_015275212.2 | XP_015130698.1 | 5 - 6 | 2030 | 65 | ||
| XM_015275219.2 | XP_015130705.1 | 5 - 6 | 2030 | 65 | ||
| XM_015275226.2 | XP_015130712.1 | 4 - 5 | 1874 | 65 | ||
| XM_015275228.2 | XP_015130714.1 | 4 - 5 | 1156 | 65 | ||
| XM_015275234.2 | XP_015130720.1 | 3 - 4 | 1712 | 65 | ||
| GenBank mRNA | AF427472.1 | - | 4 - 5 | 1048 | 65 | |
More Information
Set Membership: |
|

DACH1
Genomic Map