Assay Details
| Gene Symbol: TAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135968.1 | NP_001129440.1 | 1 - 2 | 101 | 70 | |
| GenBank mRNA | AJ843261.1 | - | 1 - 2 | 101 | 70 | |
| JF794477.1 | - | 1 - 2 | 101 | 70 | ||
| JF794478.1 | - | 1 - 2 | 101 | 70 | ||
| JF794479.1 | - | 1 - 2 | 101 | 70 | ||
| JF794480.1 | - | 1 - 2 | 101 | 70 | ||
| JF794481.1 | - | 1 - 2 | 101 | 70 | ||
| JF794482.1 | - | 1 - 2 | 101 | 70 | ||
| JF794483.1 | - | 1 - 2 | 70 | |||
More Information
Set Membership: |
|

TAP1
Genomic Map