Assay Details
| Gene Symbol: MRPS18A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277623.1 | NP_001264552.1 | 4 - 5 | 382 | 59 | |
| NM_001277624.1 | NP_001264553.1 | 4 - 5 | 382 | 59 | ||
| GenBank mRNA | BG709792.1 | - | 4 - 5 | 382 | 59 | |
| BU136195.1 | - | 4 - 5 | 373 | 59 | ||
| BU357038.1 | - | 4 - 5 | 375 | 59 | ||
| BX929638.1 | - | 4 - 5 | 378 | 59 | ||
| BX932269.1 | - | 4 - 5 | 376 | 59 | ||
| BX935499.1 | - | 4 - 5 | 373 | 59 | ||
More Information
Set Membership: |
|

MRPS18A
Genomic Map