Assay Details
| Gene Symbol: UGT1A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015289249.2 | XP_015144735.1 | 3 - 4 | 1106 | 89 | |
| XM_015289250.2 | XP_015144736.1 | 3 - 4 | 1134 | 89 | ||
| XM_015289251.2 | XP_015144737.2 | 3 - 4 | 1121 | 89 | ||
| XM_015289252.2 | XP_015144738.1 | 3 - 4 | 1150 | 89 | ||
| XM_015289253.1 | XP_015144739.1 | 3 - 4 | 1095 | 89 | ||
| XM_015289254.2 | XP_015144740.2 | 5 - 6 | 2271 | 89 | ||
| XM_015289255.2 | XP_015144741.1 | 3 - 4 | 1155 | 89 | ||
| XM_015289256.2 | XP_015144742.1 | 3 - 4 | 1211 | 89 | ||
| XM_025152657.1 | XP_025008425.1 | 4 - 5 | 2195 | 89 | ||
| GenBank mRNA | BX936208.2 | - | 3 - 4 | 733 | 89 | |
More Information
Set Membership: |
|

UGT1A1
Genomic Map