Assay Details
Gene Symbol: BEND7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001282235.2 | NP_001269164.1 | 4 - 5 | 839 | 64 | |
GenBank mRNA | CR388896.1 | - | 4 - 5 | 840 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
