Assay Details
| Gene Symbol: KLHL34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004934669.3 | XP_004934726.1 | 1 | 3007 | 102 | |
| GenBank mRNA | CR389218.1 | - | 1 | 548 | 102 | |
More Information
Set Membership: |
|

KLHL34
Genomic Map