Assay Details
| Gene Symbol: ENDOG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004945854.3 | XP_004945911.1 | 1 - 2 | 587 | 68 | |
| XM_415487.6 | XP_415487.2 | 1 - 2 | 572 | 68 | ||
| GenBank mRNA | CR389245.1 | - | 1 - 2 | 174 | 68 | |
More Information
Set Membership: |
|

ENDOG
Genomic Map