Assay Details
| Gene Symbol: USF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277825.1 | NP_001264754.1 | 5 - 6 | 365 | 122 | |
| GenBank mRNA | AM070924.1 | - | 5 - 6 | 365 | 122 | |
| CR390051.1 | - | 1 - 2 | 93 | 122 | ||
More Information
Set Membership: |
|

USF3
Genomic Map