Assay Details
| Gene Symbol: ETAA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004940211.3 | XP_004940268.1 | 5 - 6 | 2912 | 156 | |
| GenBank mRNA | CR390218.1 | - | 1 - 2 | 405 | 156 | |
More Information
Set Membership: |
|

ETAA1
Genomic Map