Assay Details
| Gene Symbol: NMU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277921.1 | NP_001264850.1 | 2 - 3 | 221 | 78 | |
| NM_001277922.1 | NP_001264851.1 | 2 - 3 | 221 | 78 | ||
| GenBank mRNA | BM491286.1 | - | 2 - 3 | 221 | 78 | |
| BU278047.1 | - | 2 - 3 | 86 | 78 | ||
| BU298768.1 | - | 2 - 3 | 211 | 78 | ||
| CR390466.1 | - | 2 - 3 | 129 | 78 | ||
| MF967031.1 | - | 2 - 3 | 235 | 78 | ||
More Information
Set Membership: |
|

NMU
Genomic Map