Assay Details
| Gene Symbol: UQCRQ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_414651.4 | XP_414651.2 | 1 - 2 | 220 | 70 | |
| GenBank mRNA | BX933197.1 | - | 1 - 2 | 200 | 70 | |
| CR405815.1 | - | 1 - 2 | 165 | 70 | ||
More Information
Set Membership: |
|

UQCRQ
Genomic Map