Assay Details
Gene Symbol: FKBP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_421981.6 | XP_421981.6 | 1 - 2 | 539 | 73 | |
GenBank mRNA | CR353563.1 | - | 1 - 2 | 430 | 73 | |
CR405926.1 | - | 1 - 2 | 235 | 73 | ||
JQ004983.1 | - | 1 - 2 | 245 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
