Assay Details
| Gene Symbol: LOC422821 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | CR407294.1 | - | 1 | 552 | 118 | |
| Gene Symbol: LCORL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025149698.1 | XP_025005466.1 | 7 | 1978 | 118 | |
More Information
Set Membership: |
|

LOC422821
Genomic Map