Assay Details
| Gene Symbol: AGT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004935493.3 | XP_004935550.1 | 4 - 5 | 1171 | 92 | |
| XM_004935494.3 | XP_004935551.1 | 3 - 4 | 1048 | 92 | ||
| XM_025149147.1 | XP_025004915.1 | 2 - 3 | 990 | 92 | ||
| XM_419584.6 | XP_419584.3 | 2 - 3 | 896 | 92 | ||
| GenBank mRNA | BX950558.1 | - | 2 - 3 | 867 | 92 | |
| CR407474.1 | - | 2 - 3 | 907 | 92 | ||
More Information
Set Membership: |
|

AGT
Genomic Map