Assay Details
| Gene Symbol: MRPS35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278091.1 | NP_001265020.1 | 2 - 3 | 281 | 66 | |
| NM_001278092.1 | NP_001265021.1 | 2 - 3 | 223 | 66 | ||
| GenBank mRNA | BX935160.1 | - | 2 - 3 | 215 | 66 | |
| CR522886.1 | - | 2 - 3 | 113 | 66 | ||
| DT655029.1 | - | 2 - 3 | 288 | 66 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001278092.1 transcripts.
Set Membership: |
|

MRPS35
Genomic Map