Assay Details
| Gene Symbol: CALCA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113708.1 | NP_001107180.1 | 2 - 3 | 166 | 76 | |
| NM_001271964.1 | NP_001258893.1 | 2 - 3 | 235 | 76 | ||
| NM_001271965.1 | NP_001258894.1 | 2 - 3 | 235 | 76 | ||
| NM_001271966.1 | NP_001258895.1 | 2 - 3 | 235 | 76 | ||
| XM_015286339.2 | XP_015141825.1 | 2 - 3 | 232 | 76 | ||
| XM_015286340.2 | XP_015141826.1 | 2 - 3 | 172 | 76 | ||
| XM_025150612.1 | XP_025006380.1 | 9 - 10 | 2798 | 76 | ||
| GenBank mRNA | BM490287.1 | - | 2 - 3 | 157 | 76 | |
| BU203257.1 | - | 2 - 3 | 238 | 77 | ||
| BU213401.1 | - | 2 - 3 | 234 | 76 | ||
| BU262345.1 | - | 2 - 3 | 232 | 76 | ||
| BU335410.1 | - | 2 - 3 | 131 | 76 | ||
| EU367492.1 | - | 1 - 2 | 92 | 76 | ||
| X03012.1 | - | 59 | 76 | |||
More Information
Set Membership: |
|

CALCA
Genomic Map