Assay Details
| Gene Symbol: C14H16orf59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_036657.2 | 3 - 4 | 657 | 84 | ||
| GenBank mRNA | BX930203.1 | - | 3 - 4 | 279 | 84 | |
| BX930271.1 | - | 3 - 4 | 657 | 84 | ||
| BX932942.1 | - | 2 - 3 | 293 | 84 | ||
| BX933532.2 | - | 2 - 3 | 221 | 84 | ||
| BX934307.2 | - | 3 - 4 | 172 | 84 | ||
| BX934976.2 | - | 3 - 4 | 666 | 84 | ||
More Information
Set Membership: |
|

C14H16orf59
Genomic Map