Assay Details
| Gene Symbol: C14H16orf59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BX930203.1 | - | 4 - 5 | 373 | 104 | |
| BX932634.1 | - | 2 - 3 | 137 | 104 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C14H16orf59| Gene Symbol: C14H16orf59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BX930203.1 | - | 4 - 5 | 373 | 104 | |
| BX932634.1 | - | 2 - 3 | 137 | 104 | ||
Set Membership: |
|