Assay Details
Gene Symbol: NINJ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277877.1 | NP_001264806.1 | 1 - 2 | 130 | 125 | |
GenBank mRNA | BX930736.1 | - | 1 - 2 | 231 | 125 | |
BX935591.1 | - | 1 - 2 | 130 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
