Assay Details
| Gene Symbol: MRPL19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025149291.1 | XP_025005059.1 | 4 - 5 | 516 | 89 | |
| XM_420071.6 | XP_420071.5 | 4 - 5 | 453 | 89 | ||
| GenBank mRNA | BX930993.1 | - | 4 - 5 | 446 | 89 | |
| BX932025.1 | - | 3 - 4 | 484 | 89 | ||
| BX932217.2 | - | 4 - 5 | 436 | 89 | ||
| BX934005.1 | - | 4 - 5 | 409 | 89 | ||
More Information
Set Membership: |
|

MRPL19
Genomic Map