Assay Details
| Gene Symbol: CA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_415893.6 | XP_415893.1 | 2 - 3 | 160 | 76 | |
| GenBank mRNA | BX931094.1 | - | 1 - 2 | 44 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CA4| Gene Symbol: CA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_415893.6 | XP_415893.1 | 2 - 3 | 160 | 76 | |
| GenBank mRNA | BX931094.1 | - | 1 - 2 | 44 | 76 | |
Set Membership: |
|