Assay Details
| Gene Symbol: NCBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277449.2 | NP_001264378.1 | 1 - 2 | 153 | 63 | |
| NM_001277455.1 | NP_001264384.1 | 1 - 2 | 153 | 63 | ||
| XM_025153360.1 | XP_025009128.1 | 1 - 2 | 1394 | 63 | ||
| GenBank mRNA | BX932072.1 | - | 1 - 2 | 150 | 63 | |
| BX934003.2 | - | 1 - 2 | 124 | 63 | ||
| CD733179.1 | - | 1 - 2 | 154 | 63 | ||
| CN220291.1 | - | 1 - 2 | 132 | 63 | ||
| DR417660.1 | - | 192 | 63 | |||
More Information
Set Membership: |
|

NCBP2
Genomic Map