Assay Details
| Gene Symbol: EMC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001234670.5 | XP_001234671.1 | 2 - 3 | 142 | 93 | |
| GenBank mRNA | BX933767.1 | - | 2 - 3 | 134 | 93 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001234670.5 transcripts.
Set Membership: |
|

EMC6
Genomic Map