Assay Details
| Gene Symbol: PAPOLB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198601.3 | NP_001185530.1 | 8 - 9 | 971 | 128 | |
| XM_015287288.2 | XP_015142774.1 | 8 - 9 | 978 | 128 | ||
| XM_015287290.2 | XP_015142776.1 | 8 - 9 | 978 | 128 | ||
| XM_015287291.2 | XP_015142777.1 | 8 - 9 | 978 | 128 | ||
| GenBank mRNA | AF061758.1 | - | 4 - 5 | 344 | 128 | |
| BX934124.1 | - | 8 - 9 | 743 | 128 | ||
| BX934578.2 | - | 7 - 8 | 793 | 128 | ||
More Information
Set Membership: |
|

PAPOLB
Genomic Map