Assay Details
Gene Symbol: IDS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277660.1 | NP_001264589.1 | 2 - 3 | 254 | 69 | |
XM_015278303.2 | XP_015133789.2 | 3 - 4 | 286 | 69 | ||
XM_015278304.2 | XP_015133790.1 | 3 - 4 | 402 | 69 | ||
XM_015278305.2 | XP_015133791.1 | 3 - 4 | 389 | 69 | ||
XM_015278306.2 | XP_015133792.1 | 2 - 3 | 320 | 69 | ||
XM_015278307.2 | XP_015133793.1 | 3 - 4 | 368 | 69 | ||
XM_015278308.2 | XP_015133794.1 | 3 - 4 | 270 | 69 | ||
XM_025149575.1 | XP_025005343.1 | 3 - 4 | 359 | 69 | ||
XM_025149576.1 | XP_025005344.1 | 4 - 5 | 358 | 69 | ||
XM_025149577.1 | XP_025005345.1 | 4 - 5 | 373 | 69 | ||
XM_025149578.1 | XP_025005346.1 | 2 - 3 | 260 | 69 | ||
GenBank mRNA | BU247591.1 | - | 2 - 3 | 245 | 69 | |
BX934826.1 | - | 2 - 3 | 246 | 69 | ||
CD215596.1 | - | 2 - 3 | 250 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
