Assay Details
| Gene Symbol: CNRIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277530.1 | NP_001264459.1 | 2 - 3 | 422 | 122 | |
| GenBank mRNA | BX935010.1 | - | 2 - 3 | 422 | 122 | |
| DN855830.1 | - | 2 - 3 | 344 | 122 | ||
More Information
Set Membership: |
|

CNRIP1
Genomic Map