Assay Details
Gene Symbol: TXNL4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278071.1 | NP_001265000.1 | 2 - 3 | 323 | 106 | |
NM_001278072.1 | NP_001265001.1 | 1 - 2 | 356 | 106 | ||
XM_015296138.1 | XP_015151624.1 | 2 - 3 | 259 | 106 | ||
GenBank mRNA | BU424492.1 | - | 1 - 2 | 358 | 106 | |
BX930428.2 | - | 1 - 2 | 287 | 106 | ||
BX931530.2 | - | 563 | 106 | |||
BX935128.1 | - | 1 - 2 | 350 | 106 | ||
CO770541.1 | - | 2 - 3 | 406 | 106 | ||
CR354018.1 | - | 1 - 2 | 287 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
