Assay Details
Gene Symbol: FAM103A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277107.1 | NP_001264036.1 | 2 - 3 | 348 | 81 | |
NM_001277108.1 | NP_001264037.1 | 2 - 3 | 228 | 81 | ||
XM_015291993.2 | XP_015147479.1 | 2 - 3 | 250 | 81 | ||
XM_015291994.1 | XP_015147480.1 | 2 - 3 | 245 | 81 | ||
GenBank mRNA | BX930464.1 | - | 2 - 3 | 228 | 81 | |
BX935339.1 | - | 2 - 3 | 205 | 81 | ||
BX935555.1 | - | 2 - 3 | 219 | 81 | ||
BX936107.1 | - | 2 - 3 | 199 | 81 | ||
CD731131.1 | - | 2 - 3 | 348 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
