Assay Details
| Gene Symbol: DUPD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168006.1 | NP_001161478.1 | 3 - 4 | 472 | 103 | |
| GenBank mRNA | BX935843.1 | - | 3 - 4 | 478 | 103 | |
More Information
Set Membership: |
|

DUPD1
Genomic Map