Assay Details
| Gene Symbol: AHSA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_421292.5 | XP_421292.1 | 7 - 8 | 858 | 78 | |
| GenBank mRNA | BX935859.1 | - | 7 - 8 | 796 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AHSA1| Gene Symbol: AHSA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_421292.5 | XP_421292.1 | 7 - 8 | 858 | 78 | |
| GenBank mRNA | BX935859.1 | - | 7 - 8 | 796 | 78 | |
Set Membership: |
|