Assay Details
| Gene Symbol: PERPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277703.1 | NP_001264632.1 | 1 - 2 | 313 | 79 | |
| GenBank mRNA | BX262976.3 | - | 1 - 2 | 98 | 79 | |
| BX933061.2 | - | 1 - 2 | 279 | 79 | ||
| BX935976.1 | - | 1 - 2 | 313 | 79 | ||
More Information
Set Membership: |
|

PERPA
Genomic Map