Assay Details
| Gene Symbol: LRIF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_419232.6 | XP_419232.3 | 1923 | 101 | ||
| GenBank mRNA | BX931725.2 | - | 292 | 101 | ||
| BX950759.1 | - | 313 | 101 | |||
More Information
Set Membership: |
|

LRIF1
Genomic Map