Assay Details
| Gene Symbol: NDUFB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001302167.1 | NP_001289096.1 | 2 - 3 | 419 | 119 | |
| XM_015289421.2 | XP_015144907.1 | 2 - 3 | 250 | 119 | ||
| XM_025152183.1 | XP_025007951.1 | 2 - 3 | 195 | 119 | ||
| XM_025152184.1 | XP_025007952.1 | 2 - 3 | 254 | 119 | ||
| GenBank mRNA | BX929464.1 | - | 2 - 3 | 316 | 119 | |
| BX933072.1 | - | 1 - 2 | 775 | 119 | ||
| BX935929.1 | - | 2 - 3 | 422 | 119 | ||
| CR338759.1 | - | 2 - 3 | 419 | 119 | ||
More Information
Set Membership: |
|

NDUFB3
Genomic Map