Assay Details
Gene Symbol: CA3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277410.1 | NP_001264339.1 | 6 - 7 | 810 | 114 | |
GenBank mRNA | CR352666.1 | - | 6 - 7 | 749 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
