Assay Details
| Gene Symbol: PTRHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_420022.6 | XP_420022.3 | 1 - 2 | 266 | 89 | |
| GenBank mRNA | CR352848.1 | - | 1 - 2 | 199 | 89 | |
More Information
Set Membership: |
|

PTRHD1
Genomic Map