Assay Details
| Gene Symbol: TTC33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001232650.4 | XP_001232651.1 | 3 - 4 | 518 | 81 | |
| XM_004937213.3 | XP_004937270.1 | 3 - 4 | 354 | 81 | ||
| XM_424771.6 | XP_424771.1 | 3 - 4 | 364 | 81 | ||
| GenBank mRNA | CR353107.1 | - | 3 - 4 | 341 | 81 | |
More Information
Set Membership: |
|

TTC33
Genomic Map