Assay Details
| Gene Symbol: COX16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001197057.1 | NP_001183986.1 | 3 - 4 | 258 | 73 | |
| XM_025150715.1 | XP_025006483.1 | 3 - 4 | 534 | 73 | ||
| GenBank mRNA | BX932336.1 | - | 4 - 5 | 432 | 73 | |
| CR353996.1 | - | 4 - 5 | 434 | 73 | ||
More Information
Set Membership: |
|

COX16
Genomic Map