Assay Details
| Gene Symbol: HNMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277873.1 | NP_001264802.1 | 4 - 5 | 399 | 67 | |
| XM_015289832.2 | XP_015145318.1 | 4 - 5 | 334 | 67 | ||
| GenBank mRNA | BX929352.2 | - | 4 - 5 | 385 | 67 | |
| BX929661.2 | - | 4 - 5 | 371 | 67 | ||
| BX932177.1 | - | 5 - 6 | 286 | 67 | ||
More Information
Set Membership: |
|

HNMT
Genomic Map