Assay Details
Gene Symbol: TCTEX1D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_422724.5 | XP_422724.1 | 1 - 2 | 117 | 80 | |
GenBank mRNA | BX932411.2 | - | 1 - 2 | 136 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
