Assay Details
Gene Symbol: TAF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277796.1 | NP_001264725.1 | 1 - 2 | 48 | 74 | |
GenBank mRNA | BX932803.2 | - | 1 - 2 | 37 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
