Assay Details
Gene Symbol: TAF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277796.1 | NP_001264725.1 | 2 - 3 | 135 | 71 | |
GenBank mRNA | BU118655.1 | - | 2 - 3 | 135 | 71 | |
BU135709.1 | - | 2 - 3 | 109 | 71 | ||
BX932803.2 | - | 2 - 3 | 124 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
