Assay Details
Gene Symbol: TAF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277796.1 | NP_001264725.1 | 3 - 4 | 276 | 62 | |
GenBank mRNA | BU118655.1 | - | 3 - 4 | 276 | 62 | |
BU135709.1 | - | 3 - 4 | 250 | 62 | ||
BX932803.2 | - | 3 - 4 | 265 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
