Assay Details
Gene Symbol: TAF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277796.1 | NP_001264725.1 | 4 - 5 | 404 | 68 | |
GenBank mRNA | BU118655.1 | - | 4 - 5 | 404 | 68 | |
BU135709.1 | - | 4 - 5 | 378 | 68 | ||
BX932803.2 | - | 4 - 5 | 393 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
