Assay Details
Gene Symbol: TAF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277796.1 | NP_001264725.1 | 5 - 6 | 486 | 87 | |
GenBank mRNA | BU118655.1 | - | 5 - 6 | 486 | 87 | |
BU135709.1 | - | 5 - 6 | 460 | 87 | ||
BX932803.2 | - | 5 - 6 | 475 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
